Assay Details
Target Gene Details
Entrez Gene ID: | 243979 |
Gene Name: | MAS-related GPR, member B2 |
Gene Aliases: |
4833406I20Rik, Mgrg14 |
Location: |
Chr.7:48550965-48658094 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprb2 | NM_175531.4 | 2 | 1070 | NP_780740.2 |
| AK029369.1 | 2 | 1071 | BAC26422.1 | |
| BC107390.1 | 2 | 1068 | AAI07391.1 | |
| BC107391.2 | 2 | 1068 | AAI07392.1 |
More Information
Set Membership: |
|

Genomic Map