Assay Details
Target Gene Details
Entrez Gene ID: | 243979 |
Gene Name: | MAS-related GPR, member B2 |
Gene Aliases: |
4833406I20Rik, Mgrg14 |
Location: |
Chr.7:48550965-48658094 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprb2 |
Target Gene Details
Entrez Gene ID: | 404238 |
Gene Name: | MAS-related GPR, member B3 |
Gene Aliases: |
MrgB3 |
Location: |
Chr.7:48642863-48643801 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprb3 | NM_207537.1 | 1 | 312 | NP_997420.1 |
| BC107192.1 | 1 | 311 | AAI07193.1 | |
| BC107193.1 | 1 | 311 | AAI07194.1 | |
| BC114400.1 | 1 | 311 | AAI14401.1 | |
| BC114428.1 | 1 | 311 | AAI14429.1 |
More Information
Set Membership: |
|

Genomic Map