Assay Details
Target Gene Details
Entrez Gene ID: | 67379 |
Gene Name: | death effector domain-containing DNA binding protein 2 |
Gene Aliases: |
2410050E11Rik, FLAME-3 |
Location: |
Chr.7:25202838-25219873 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dedd2 | NM_207677.3 | 5 | 1462 | NP_997560.3 |
| AK010701.1 | 5 | 1274 | ||
| AK089096.1 | 3 | 834 | ||
| BC092250.1 | 5 | 1462 | AAH92250.1 | |
| BE956897.1 | 1 | 386 |

Genomic Map