Assay Details
Target Gene Details
Entrez Gene ID: | 243813 |
Gene Name: | leukocyte receptor cluster (LRC) member 9 |
Gene Aliases: |
9530024C23Rik, F630035L11Rik |
Location: |
Chr.7:4148183-4149872 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Leng9 | NM_175529.3 | 1 | 1123 | NP_780738.1 |
| AK089196.1 | 1 | 1123 | BAC40788.1 | |
| AK090365.1 | 1 | 740 | BAC41185.1 | |
| AK134559.1 | 1 | 744 | BAE22187.1 | |
| AK164490.1 | 1 | 1082 | ||
| BC116676.1 | 1 | 1018 |

Genomic Map