Assay Details
Target Gene Details
Entrez Gene ID: | 404240 |
Gene Name: | MAS-related GPR, member B8 |
Gene Aliases: |
MrgB8 |
Location: |
Chr.7:48388526-48389648 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprb8 | NM_207539.2 | 1 | 386 | NP_997422.1 |
| BC138164.1 | 1 | 386 | ||
| BC138165.1 | 1 | 386 |
More Information
Set Membership: |
|

Genomic Map