Assay Details
Target Gene Details
Entrez Gene ID: | 211578 |
Gene Name: | MAS-related GPR, member D |
Gene Aliases: |
Gm499, Mgrd, MrgD, TGR7 |
Location: |
Chr.7:145314835-145324059 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprd | NM_203490.3 | 1 | 77 | NP_987075.1 |
| AY762266.1 | 1 | 77 | AAV30216.1 | |
| BC137948.1 | ||||
| BC137950.1 |
More Information
Set Membership: |
|

Genomic Map