Assay Details
Target Gene Details
Entrez Gene ID: | 244238 |
Gene Name: | MAS-related GPR, member E |
Gene Aliases: |
C130069N09Rik, Ebrt3, Mgrf, MrgE |
Location: |
Chr.7:143778363-143784500 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgpre | NM_175534.3 | 2 | 898 | NP_780743.1 |
| AJ320231.1 | 2 | 609 | CAC86257.1 | |
| AK048528.1 | 2 | 716 | BAC33361.1 | |
| AK133657.1 | 2 | 900 | BAE21769.1 | |
| AK154886.1 | 2 | 693 | BAE32903.1 | |
| BC096766.1 | 2 | 694 | AAH96766.1 | |
| BC138305.1 | 2 | 607 | ||
| BC138308.1 | 2 | 607 |
More Information
Set Membership: |
|

Genomic Map