Assay Details
Target Gene Details
Entrez Gene ID: | 211577 |
Gene Name: | MAS-related GPR, member F |
Gene Aliases: |
BC019711, Mgrc, MrgF |
Location: |
Chr.7:145300909-145309557 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprf | NM_145379.2 | 3 | 1104 | NP_663354.1 |
| AK157988.1 | 4 | 1243 | BAE34302.1 | |
| BC019711.1 | 3 | 1104 | AAH19711.1 |
More Information
Set Membership: |
|

Genomic Map