Assay Details
Target Gene Details
Entrez Gene ID: | 404242 |
Gene Name: | MAS-related GPR, member X1 |
Gene Aliases: |
MrgC11, Mrgprc11 |
Location: |
Chr.7:48020971-48027597 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprx1 | NM_207540.3 | 2 | 1009 | NP_997423.1 |
| AY152435.1 | 2 | 1015 | AAN64385.1 | |
| BC150669.1 | 2 | 956 |
More Information
Set Membership: |
|

Genomic Map