Assay Details
Target Gene Details
Entrez Gene ID: | 243978 |
Gene Name: | MAS-related GPR, member X2 |
Gene Aliases: |
G370024M05Rik, MrgB10, Mrgprb10 |
Location: |
Chr.7:48478619-48499270 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprx2 | NM_001034868.3 | 2 | 872 | NP_001030040.2 |
| AK148121.1 | 2 | 803 | BAE28359.1 | |
| AK148153.1 | BAE28379.1 | |||
| AK148319.1 | 1 | 193 | BAE28478.1 | |
| BC141225.1 | 2 | 795 |
More Information
Set Membership: |
|

Genomic Map