Assay Details
Target Gene Details
Entrez Gene ID: | 66126 |
Gene Name: | elongation factor 1 homolog (ELF1, S. cerevisiae) |
Gene Aliases: |
1110011K10Rik |
Location: |
Chr.9:22112989-22114169 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elof1 | NM_170777.3 | 2 | 828 | NP_740747.1 |
| BC056225.1 | 2 | 828 | AAH56225.1 |

Genomic Map