Assay Details
Target Gene Details
Entrez Gene ID: | 320844 |
Gene Name: | adhesion molecule with Ig like domain 3 |
Gene Aliases: |
E430002N15Rik, ali3, mKIAA1851 |
Location: |
Chr.9:108053159-108055701 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amigo3 | NM_177275.4 | 1 | 1689 | NP_796249.1 |
| AB167510.1 | 1 | 1637 | BAD12542.1 | |
| AK028619.1 | 1 | 1637 | BAC26035.1 | |
| AK088051.1 | 1 | 1690 | BAC40120.1 | |
| AY237004.1 | 1 | 1468 | AAO48945.1 |
Target Gene Details
Entrez Gene ID: | 84585 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
BC003945, Kpc1 |
Location: |
Chr.9:108051672-108079773 on Build GRCm38 |
Assay Gene Location: | Within Intron 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf123 | NM_032543.2 | NP_115932.1 | ||
| AK137599.1 | ||||
| AK172244.1 | ||||
| AK212142.1 | ||||
| AK216931.1 | ||||
| AY744153.1 | AAU93471.1 | |||
| BC025530.1 | ||||
| BC057082.1 | AAH57082.1 |
More Information
Set Membership: |
|

Genomic Map