Assay Details
Target Gene Details
Entrez Gene ID: | 72454 |
Gene Name: | coiled-coil domain containing 71 |
Gene Aliases: |
2600016J21Rik |
Location: |
Chr.9:108452571-108465945 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc71 | NM_133744.4 | 2 | 1608 | NP_598505.1 |
| AK011229.1 | 2 | 1574 | ||
| AK044601.1 | 2 | 1574 | BAC31997.1 | |
| AK079333.1 | 2 | 1581 | ||
| AK085544.1 | 2 | 1600 | ||
| AK086560.1 | 2 | 1574 | BAC39691.1 | |
| AK136982.1 | 2 | 1561 | ||
| AK149878.1 | 2 | 1574 | BAE29140.1 | |
| AK161396.1 | 2 | 1684 | BAE36372.1 | |
| BC018518.1 | 2 | 1559 | AAH18518.1 |

Genomic Map