Assay Details
Target Gene Details
Entrez Gene ID: | 70359 |
Gene Name: | GTP binding protein 3 |
Gene Aliases: |
2410009F13Rik, AI607903 |
Location: |
Chr.8:71487870-71499623 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp3 | NM_032544.3 | 8 | 1834 | NP_115933.2 |
| AF361482.2 | 8 | 1834 | AAK35216.2 | |
| AK010459.1 | 1 | 144 | BAC25298.1 | |
| AK046330.1 | 8 | 1210 | ||
| BC092042.1 | 7 | 1144 | AAH92042.1 | |
| BC138560.1 | 8 | 1243 |
More Information
Set Membership: |
|

Genomic Map