Assay Details
Target Gene Details
Entrez Gene ID: | 70726 |
Gene Name: | angiopoietin-like 6 |
Gene Aliases: |
6330404E11Rik, AGF, ARP3, Arp5 |
Location: |
Chr.9:20873808-20879816 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angptl6 | NM_145154.2 | 5 | 1127 | NP_660136.1 |
| AB054065.1 | 5 | 1127 | BAB91249.1 | |
| AK018113.1 | 4 | 496 | ||
| AK218362.1 | 2 | 221 | ||
| BC025904.1 | 5 | 1105 | AAH25904.1 |
More Information
Set Membership: |
|

Genomic Map