Assay Details
Target Gene Details
Entrez Gene ID: | 319278 |
Gene Name: | RIKEN cDNA A230050P20 gene |
Gene Aliases: |
- |
Location: |
Chr.9:20868592-20874307 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A230050P20Rik | NM_175687.2 | 8 | 1270 | NP_783618.1 |
| AK034516.1 | 7 | 2720 | ||
| AK149161.1 | 8 | 1250 | BAE28753.1 | |
| AK157146.1 | 7 | 2798 | BAE33979.1 | |
| BC031510.1 | 7 | 1420 | ||
| BC043448.1 | 8 | 1216 | AAH43448.1 | |
| BY625900.1 |
Target Gene Details
Entrez Gene ID: | 70726 |
Gene Name: | angiopoietin-like 6 |
Gene Aliases: |
6330404E11Rik, AGF, ARP3, Arp5 |
Location: |
Chr.9:20873808-20879816 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angptl6 | NM_145154.2 | 6 | 1479 | NP_660136.1 |
| AB054065.1 | 6 | 1479 | BAB91249.1 | |
| AK018113.1 | ||||
| BC025904.1 | 6 | 1457 | AAH25904.1 |
More Information
Set Membership: |
|

Genomic Map