Assay Details
Target Gene Details
Entrez Gene ID: | 108043 |
Gene Name: | cholinergic receptor, nicotinic, beta polypeptide 3 |
Gene Aliases: |
5730417K16Rik, Acrb3 |
Location: |
Chr.8:27368711-27399730 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrnb3 | NM_027454.4 | 1 | 51 | NP_081730.1 |
| NM_173212.4 | 1 | 51 | NP_775304.1 | |
| AK017571.1 | BAB30812.1 | |||
| AK030464.1 | 1 | 52 | BAC26973.1 | |
| AK133689.1 | BAE21785.1 |
More Information
Set Membership: |
|

Genomic Map