Assay Details
Target Gene Details
Entrez Gene ID: | 17388 |
Gene Name: | matrix metallopeptidase 15 |
Gene Aliases: |
AI503551, MT2-MMP |
Location: |
Chr.8:95352337-95374293 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp15 | NM_008609.3 | 1 | 333 | NP_032635.1 |
| BC047278.1 | 1 | 333 | AAH47278.1 |
More Information
Set Membership: |
|

Genomic Map