Assay Details
Target Gene Details
Entrez Gene ID: | 56513 |
Gene Name: | par-6 family cell polarity regulator alpha |
Gene Aliases: |
0710008C04Rik, 2610010A15Rik, PAR-6A, PAR6alpha, Par-6, Par6, Par6c, TAX40, Tip-40 |
Location: |
Chr.8:105701148-105703494 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pard6a | NM_001047435.2 | 3 | 972 | NP_001040900.1 |
| NM_001047436.2 | 3 | 988 | NP_001040901.1 | |
| NM_001286344.1 | 3 | 957 | NP_001273273.1 | |
| NM_001286345.1 | 3 | 885 | NP_001273274.1 | |
| NM_019695.3 | 3 | 975 | NP_062669.2 | |
| AF070970.1 | 3 | 971 | AAD15928.1 | |
| AK003030.1 | 3 | 941 | ||
| AK158848.1 | 3 | 970 | BAE34693.1 | |
| AK209727.1 | 1 | 210 | ||
| BC049593.1 | 3 | 949 | AAH49593.1 | |
| BC061114.1 | 3 | 885 | AAH61114.1 | |
| BF466814.1 | 1 | 310 | ||
| CO040458.1 | 1 | 310 |
More Information
Set Membership: |
|

Genomic Map