Assay Details
Target Gene Details
Entrez Gene ID: | 68861 |
Gene Name: | RIKEN cDNA 1190002N15 gene |
Gene Aliases: |
GoPro49, HASF |
Location: |
Chr.9:94517864-94538081 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1190002N15Rik | NM_001033145.2 | 2 | 1025 | NP_001028317.2 |
| AK162197.1 | 1 | 149 | BAE36786.1 |

Genomic Map