Assay Details
Target Gene Details
Entrez Gene ID: | 330998 |
Gene Name: | ankyrin repeat domain 34C |
Gene Aliases: |
B230218L05Rik, BB276295, Gm771 |
Location: |
Chr.9:89723933-89738877 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd34c | NM_207260.2 | 1 | 334 | NP_997143.1 |
| AK045652.1 | 1 | 336 | BAC32445.1 | |
| BC137642.1 | ||||
| BC137643.1 |

Genomic Map