Assay Details
Target Gene Details
Entrez Gene ID: | 12500 |
Gene Name: | CD3 antigen, delta polypeptide |
Gene Aliases: |
T3d |
Location: |
Chr.9:44981769-44987052 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd3d | NM_013487.3 | 5 | 940 | NP_038515.3 |
| AK042172.1 | 5 | 942 | BAE20624.1 | |
| AK154064.1 | 5 | 902 | BAE32350.1 | |
| BY576367.1 |

Genomic Map