Assay Details
Target Gene Details
Entrez Gene ID: | 64933 |
Gene Name: | adaptor-related protein complex 3, mu 2 subunit |
Gene Aliases: |
5830445E16Rik, AP-3B |
Location: |
Chr.8:22787354-22805654 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap3m2 | NM_001122820.1 | 9 | 2072 | NP_001116292.1 |
| NM_029505.3 | 9 | 2010 | NP_083781.2 | |
| AK081220.1 | 9 | 1979 | BAC38169.1 | |
| AK082988.1 | 9 | 1968 | BAC38723.1 | |
| AK134634.1 | 9 | 2022 | BAE22219.1 | |
| AK162771.1 | 4 | 1115 | BAE37054.1 | |
| AY039763.1 | 9 | 1990 | AAK73278.1 | |
| BC027301.1 | 9 | 1956 | AAH27301.1 | |
| BC030484.1 | 9 | 1994 | AAH30484.1 |
More Information
Set Membership: |
|

Genomic Map