Assay Details
Target Gene Details
Entrez Gene ID: | 619331 |
Gene Name: | zinc fingr protein 551 |
Gene Aliases: |
9630004E07Rik, Znf551 |
Location: |
Chr.7:12412548-12422739 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp551 | NM_001033820.3 | 4 | 954 | NP_001028992.2 |
| AK137311.1 | 4 | 955 | BAE23302.1 | |
| BC141160.1 | 4 | 1052 |

Genomic Map