Assay Details
Target Gene Details
Entrez Gene ID: | 68196 |
Gene Name: | heat shock factor binding protein 1 |
Gene Aliases: |
0610007A03Rik, AA408769, Hsp25 |
Location: |
Chr.8:119344538-119348929 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsbp1 | NM_024219.1 | 4 | 424 | NP_077181.1 |
| AK018708.1 | 4 | 456 | BAB31359.1 | |
| AK050550.1 | 4 | 474 | BAC34319.1 | |
| AK078498.1 | 4 | 456 | BAC37310.1 | |
| AK145940.1 | 4 | 434 | BAE26770.1 | |
| BC002153.1 | 4 | 424 | AAH02153.1 |

Genomic Map