Assay Details
Target Gene Details
Entrez Gene ID: | 270096 |
Gene Name: | MON1 homolog b (yeast) |
Gene Aliases: |
5031407H10, 5033413H12Rik |
Location: |
Chr.8:113635586-113645192 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mon1b | NM_001048143.2 | 3 | 515 | NP_001041608.1 |
| NM_173015.3 | 2 | 705 | NP_766603.1 | |
| AK030288.1 | 2 | 671 | BAC26880.1 | |
| AK157390.1 | 3 | 515 | ||
| AK173047.1 | 2 | 377 | BAD32325.1 | |
| BC062639.1 | 2 | 381 | AAH62639.1 | |
| BY742778.1 | ||||
| CA320533.1 | 2 | 295 | ||
| CJ089261.1 | 3 | 516 |

Genomic Map