Assay Details
Target Gene Details
Entrez Gene ID: | 112418 |
Gene Name: | RIKEN cDNA 1700102P08 gene |
Gene Aliases: |
AI593206 |
Location: |
Chr.9:108392834-108410966 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700102P08Rik | NM_053216.2 | 4 | 807 | NP_444446.2 |
| AI593206.1 | 1 | 307 | ||
| AK007115.1 | 4 | 807 | BAB24864.1 | |
| BC061048.1 | 4 | 807 | AAH61048.1 |

Genomic Map