Assay Details
Target Gene Details
Entrez Gene ID: | 208117 |
Gene Name: | anterior pharynx defective 1b homolog (C. elegans) |
Gene Aliases: |
2310057K14Rik, Aph1c |
Location: |
Chr.9:66775202-66797293 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aph1b | NM_177583.4 | 6 | 3261 | NP_808251.1 |
| AK028521.1 | 7 | 3528 | BAC25988.1 | |
| AK049828.1 | 6 | 3262 | BAC33940.1 | |
| AK154847.1 | 6 | 3329 | BAE32873.1 | |
| AK162585.1 | 7 | 3496 | BAE36975.1 | |
| BC079659.1 | 6 | 3293 | AAH79659.1 |
More Information
Set Membership: |
|

Genomic Map