Assay Details
Target Gene Details
Entrez Gene ID: | 68318 |
Gene Name: | anterior pharynx defective 1c homolog (C. elegans) |
Gene Aliases: |
0610008A10Rik, APH-1b, Aph1b |
Location: |
Chr.9:66814994-66834779 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aph1c | NM_026674.3 | 6 | 2954 | NP_080950.1 |
| BC050923.1 | 6 | 2954 | AAH50923.1 | |
| BC063254.1 | 6 | 2954 | AAH63254.1 |
More Information
Set Membership: |
|

Genomic Map