Assay Details
Target Gene Details
Entrez Gene ID: | 69068 |
Gene Name: | RIKEN cDNA 1810011O10 gene |
Gene Aliases: |
1110065B09Rik, AW121743, AW321058 |
Location: |
Chr.8:24437616-24438946 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810011O10Rik | NM_026931.2 | 1 | 447 | NP_081207.1 |
| AK007441.1 | 1 | 448 | BAB25041.1 | |
| BC016562.1 | 1 | 404 | AAH16562.1 |

Genomic Map