Assay Details
Target Gene Details
Entrez Gene ID: | 107566 |
Gene Name: | ADP-ribosylation factor-like 2 binding protein |
Gene Aliases: |
1700010P10Rik, 1700027H16Rik, 6330544B05Rik, AI482273, AI849834, BART, Bart1 |
Location: |
Chr.8:94666600-94674457 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl2bp | NM_024191.2 | 6 | 744 | NP_077153.1 |
| NM_024269.4 | 6 | 582 | NP_077231.1 | |
| AK006418.1 | 6 | 569 | BAB24578.1 | |
| AK006646.1 | 6 | 568 | BAB24686.1 | |
| AK018229.1 | 6 | 554 | BAB31126.1 | |
| AK149620.1 | 6 | 581 | ||
| AK150513.1 | 6 | 746 | ||
| AK155242.1 | 6 | 695 | BAE33143.1 | |
| AK161325.1 | 6 | 563 | BAE36323.1 | |
| AK181415.1 | ||||
| AK183790.1 | ||||
| BC002131.1 | 6 | 589 | AAH02131.1 | |
| BC024708.1 | 6 | 693 | AAH24708.1 | |
| CT010168.1 | CAJ18376.1 |

Genomic Map