Assay Details
Target Gene Details
Entrez Gene ID: | 102122 |
Gene Name: | family with sequence similarity 192, member A |
Gene Aliases: |
1700001O11Rik, 2310065K24Rik, AI596259, Nip30 |
Location: |
Chr.8:94574940-94601726 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam192a | NM_028221.4 | 8 | 1643 | NP_082497.2 |
| AK033988.1 | BAC28539.1 | |||
| AK050704.1 | 8 | 1578 | BAC34387.1 | |
| AK151603.1 | 8 | 1597 | BAE30543.1 | |
| BC016088.1 | 7 | 1383 | AAH16088.1 | |
| BC025472.1 | 8 | 1619 | AAH25472.1 | |
| BC027060.1 | 8 | 1605 | AAH27060.1 | |
| BC027818.1 | 7 | 1411 |

Genomic Map