Assay Details
Target Gene Details
Entrez Gene ID: | 403395 |
Gene Name: | C-type lectin domain family 3, member a |
Gene Aliases: |
1110019O10Rik, Clecsf1, Gm796 |
Location: |
Chr.8:114418034-114428133 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec3a | NM_001007223.3 | 3 | 440 | NP_001007224.1 |
| BC147139.1 | 3 | 440 | ||
| BC147140.1 | 3 | 440 |
More Information
Set Membership: |
|

Genomic Map