Assay Details
Target Gene Details
Entrez Gene ID: | 234836 |
Gene Name: | interleukin 17C |
Gene Aliases: |
IL-17C |
Location: |
Chr.8:122422085-122423639 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il17c | NM_145834.3 | 1 | 106 | NP_665833.3 |
| AF458061.1 | 1 | 106 | AAM77565.1 | |
| BC145853.1 | 1 | 169 |

Genomic Map