Assay Details
Target Gene Details
Entrez Gene ID: | 434280 |
Gene Name: | predicted gene 5607 |
Gene Aliases: |
EG434280 |
Location: |
Chr.8:12385771-12436732 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5607 | NR_027975.2 | |||
| BC066076.1 | AAH66076.1 |
Target Gene Details
Entrez Gene ID: | 20664 |
Gene Name: | SRY (sex determining region Y)-box 1 |
Gene Aliases: |
BB176347, Sox-1 |
Location: |
Chr.8:12395519-12399555 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox1 | NM_009233.3 | 1 | 3356 | NP_033259.2 |
More Information
Set Membership: |
|

Genomic Map