Assay Details
Target Gene Details
Entrez Gene ID: | 59046 |
Gene Name: | cAMP-regulated phosphoprotein 19 |
Gene Aliases: |
19kDa, 2700024H10Rik, ARPP-19, AW559096, Arpp12 |
Location: |
Chr.9:75037614-75060313 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpp19 | NM_001142655.1 | 4 | 2740 | NP_001136127.1 |
| NM_021548.4 | 3 | 2809 | NP_067523.1 | |
| AK049760.1 | 3 | 2631 | BAC33908.1 | |
| BC040206.1 | 3 | 2809 | AAH40206.1 |

Genomic Map