Assay Details
Target Gene Details
Entrez Gene ID: | 110834 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 3 |
Gene Aliases: |
(a)3, A730007P14Rik, Acra-3, Acra3 |
Location: |
Chr.9:55011343-55026559 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna3 | NM_145129.2 | 5 | 1028 | NP_660111.2 |
| AF325346.1 | 5 | 898 | AAL37362.1 | |
| AF459029.1 | 5 | 1001 | AAL58471.1 | |
| AF472588.1 | 5 | 930 | AAL84757.1 | |
| AK051730.1 | 5 | 1032 | BAC34740.1 | |
| AK053497.1 | 5 | 956 | BAC35404.1 | |
| AK080415.1 | 5 | 1029 | BAC37909.1 | |
| AK080475.1 | 5 | 1029 | ||
| AK148786.1 | 5 | 1000 | ||
| AY574262.1 | 5 | 863 | AAS90358.1 | |
| BC120597.1 | 5 | 899 | ||
| BC120599.1 | 5 | 899 |
More Information
Set Membership: |
|

Genomic Map