Assay Details
Target Gene Details
Entrez Gene ID: | 110834 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 3 |
Gene Aliases: |
(a)3, A730007P14Rik, Acra-3, Acra3 |
Location: |
Chr.9:55011343-55026559 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna3 | NM_145129.2 | 6 | 2596 | NP_660111.2 |
| AK051730.1 | 6 | 2600 | BAC34740.1 | |
| AK080415.1 | 6 | 2597 | BAC37909.1 |
More Information
Set Membership: |
|

Genomic Map