Assay Details
Target Gene Details
Entrez Gene ID: | 108015 |
Gene Name: | cholinergic receptor, nicotinic, beta polypeptide 4 |
Gene Aliases: |
Acrb-4, Acrb4 |
Location: |
Chr.9:55028154-55048947 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrnb4 | NM_148944.4 | 5 | 1236 | NP_683746.1 |
| AF325351.1 | AAL37367.1 | |||
| AF492840.1 | 5 | 1221 | AAM11659.1 | |
| AK140071.1 | 5 | 1238 | BAE24228.1 | |
| AY574269.1 | 5 | 1187 | AAS90365.1 | |
| BC138851.1 | 5 | 1235 |
More Information
Set Membership: |
|

Genomic Map