Assay Details
Target Gene Details
Entrez Gene ID: | 71955 |
Gene Name: | increased sodium tolerance 1 homolog (yeast) |
Gene Aliases: |
2400003C14Rik, AW536298, mKIAA0174 |
Location: |
Chr.8:109671321-109693294 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ist1 | NM_028018.2 | 10 | 2130 | NP_082294.1 |
| AK010262.1 | 10 | 2100 | BAB26804.1 | |
| AK050468.1 | 9 | 3430 | BAC34273.1 | |
| AK088192.1 | 10 | 2085 | BAC40200.1 | |
| AK090233.1 | 1 | 1138 | ||
| AK122215.1 | 5 | 6606 | BAC65497.1 | |
| AK152830.1 | 10 | 2097 | BAE31530.1 | |
| BC017605.1 | 10 | 2096 | AAH17605.1 | |
| BC039052.1 | 9 | 3406 | AAH39052.1 | |
| BE947178.1 | 1 | 229 |
Target Gene Details
Entrez Gene ID: | 244646 |
Gene Name: | polycystic kidney disease 1 like 3 |
Gene Aliases: |
- |
Location: |
Chr.8:109613994-109673526 on Build GRCm38 |
Assay Gene Location: | Within Intron 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkd1l3 | NM_001039700.2 | NP_001034789.2 | ||
| NM_001286454.1 | NP_001273383.1 | |||
| DQ382344.1 | ABD36562.1 | |||
| DQ382345.1 | ABD36563.1 | |||
| DQ382349.1 | ABD36567.1 |
More Information
Set Membership: |
|

Genomic Map