Assay Details
Target Gene Details
Entrez Gene ID: | 72723 |
Gene Name: | zinc finger protein 74 |
Gene Aliases: |
2810054M15Rik, KRAB8, Zfp66, Znf569, mszf21, mszf77 |
Location: |
Chr.7:29931711-29955843 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp74 | NM_178384.3 | 5 | 2625 | NP_848471.2 |
| AK012947.1 | 1 | 401 | ||
| BC050048.1 | 5 | 2625 | AAH50048.1 | |
| BM242052.2 | 1 | 374 |
More Information
Set Membership: |
|

Genomic Map