Assay Details
Target Gene Details
Entrez Gene ID: | 71186 |
Gene Name: | RIKEN cDNA 4933417D19 gene |
Gene Aliases: |
- |
Location: |
Chr.8:123235091-123242359 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933417D19Rik | NR_045849.1 | |||
| AK016841.1 |
Target Gene Details
Entrez Gene ID: | 78779 |
Gene Name: | spermatogenesis associated 2-like |
Gene Aliases: |
2610039E05Rik, AI648233, mKIAA4138 |
Location: |
Chr.8:123232255-123236245 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata2l | NM_030176.2 | 2 | 235 | NP_084452.2 |
| AK019166.1 | 1 | 560 | ||
| AK081234.1 | 2 | 235 | BAC38172.1 | |
| AK170267.1 | 1 | 535 | BAE41672.1 | |
| AK177840.1 | 1 | 69 | ||
| BC139310.1 | 1 | 170 | ||
| BC139311.1 | 1 | 170 |
More Information
Set Membership: |
|

Genomic Map