Assay Details
Target Gene Details
Entrez Gene ID: | 76687 |
Gene Name: | signal peptidase complex subunit 3 homolog (S. cerevisiae) |
Gene Aliases: |
1810011E08Rik |
Location: |
Chr.8:54520433-54530739 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spcs3 | NM_029701.1 | 5 | 1492 | NP_083977.1 |
| AK028314.1 | 5 | 1454 | BAC25873.1 | |
| AK037878.1 | 1 | 887 | ||
| AK048051.1 | 5 | 1478 | BAC33224.1 | |
| AK207069.1 | ||||
| BC054817.1 | 5 | 1457 | AAH54817.1 |
More Information
Set Membership: |
|

Genomic Map