Assay Details
Target Gene Details
Entrez Gene ID: | 76900 |
Gene Name: | single stranded DNA binding protein 4 |
Gene Aliases: |
1210002E11Rik, AW743380, Sspb4 |
Location: |
Chr.8:70597485-70608380 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssbp4 | NM_133772.2 | 17 | 1434 | NP_598533.2 |
| AA271952.1 | 1 | 197 | ||
| AK004835.1 | 16 | 1791 | ||
| AK154336.1 | 17 | 1435 | BAE32522.1 | |
| AK157387.1 | 16 | 1514 | ||
| AK205273.1 | 1 | 119 | ||
| BC002150.1 | 17 | 1393 | AAH02150.1 |
More Information
Set Membership: |
|

Genomic Map