Assay Details
Target Gene Details
Entrez Gene ID: | 64931 |
Gene Name: | IZUMO1 receptor, JUNO |
Gene Aliases: |
0910001L11Rik, C86255, Folbp3, Folr4, Juno |
Location: |
Chr.9:14885814-14903951 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Izumo1r | NM_176807.4 | 5 | 978 | NP_789777.1 |
| AK041596.1 | 5 | 912 | BAC30998.1 |

Genomic Map