Assay Details
Target Gene Details
Entrez Gene ID: | 14345 |
Gene Name: | fucosyltransferase 4 |
Gene Aliases: |
AI451562, CD15, FAL, FucT-IV, LeX, SSEA-1, Ssea1 |
Location: |
Chr.9:14748459-14752122 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fut4 | NM_010242.3 | 1 | 1834 | NP_034372.1 |
| AK041752.1 | 1 | 1834 | BAC31054.1 | |
| BC137588.1 | 1 | 1825 |
More Information
Set Membership: |
|

Genomic Map