Assay Details
Target Gene Details
Entrez Gene ID: | 67722 |
Gene Name: | actin-like 11 |
Gene Aliases: |
4921517D21Rik |
Location: |
Chr.9:107928469-107932461 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actl11 | NM_026338.3 | 1 | 663 | NP_080614.1 |
| AK014908.1 | 1 | 666 | BAB29615.1 | |
| AK014926.1 | 1 | 374 | BAB29623.1 | |
| BC115773.1 | 1 | 666 |
More Information
Set Membership: |
|

Genomic Map