Assay Details
Target Gene Details
Entrez Gene ID: | 382051 |
Gene Name: | pyruvate dehyrogenase phosphatase catalytic subunit 2 |
Gene Aliases: |
4833426J09Rik, Gm1705, mKIAA1348 |
Location: |
Chr.8:104591468-104599026 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdp2 | NM_001024606.1 | 2 | 693 | NP_001019777.1 |
| AK220385.1 | 1 | 546 | BAD90442.1 | |
| BC094946.1 | 2 | 660 | AAH94946.1 |
More Information
Set Membership: |
|

Genomic Map