Assay Details
Target Gene Details
Entrez Gene ID: | 72297 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3 |
Gene Aliases: |
2210008L19Rik |
Location: |
Chr.8:71691373-71702627 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3gnt3 | NM_028189.3 | 3 | 1383 | NP_082465.3 |
| AK144180.1 | 3 | 1355 | BAE25750.1 | |
| AY039028.1 | 2 | 1302 | AAK72481.1 | |
| BC026418.1 | 3 | 1375 | AAH26418.1 |
More Information
Set Membership: |
|

Genomic Map