Assay Details
Target Gene Details
Entrez Gene ID: | 269954 |
Gene Name: | tubulin tyrosine ligase-like family, member 13 |
Gene Aliases: |
1700111A04Rik, 4921538O11, BC024578 |
Location: |
Chr.7:80246376-80260821 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll13 | NM_177765.3 | 14 | 2406 | NP_808433.2 |
| AK029565.1 | 14 | 2405 | BAC26514.1 | |
| AM690757.1 | 13 | 2107 | ||
| BC145749.1 | 13 | 2014 |
More Information
Set Membership: |
|

Genomic Map